Learn More
Selleck Chemical LLC SB273005 5mg 205678-31-5
SDP
Supplier:  Selleck Chemical LLC S75405MG
Encompass_Preferred
SB273005 is a potent integrin inhibitor with Ki of 1.2 nM and 0.3 nM for ?v?3 receptor and ?v?5 receptor, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4920