Learn More
Medchemexpress LLC HY-101502A 100mg , SB290157 (trifluoroacetate) CAS:1140525-25-2 Purity:98%
SDP
Catalog No. 501493318
Encompass_Preferred
Medchemexpress, HY-101502A 100mg SB290157 (trifluoroacetate) CAS:1140525-25-2 Formula:C24H29F3N4O6 IC50: 200 nM (C3a) Purity:98% SB290157 trifluoroacetate is a potent and selective C3a receptor antagonist with an IC 50 of 200 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3318 Supplier Medchemexpress LLC Supplier No. HY101502A100MG
Medchemexpress, HY-101502A 100mg SB290157 (trifluoroacetate) CAS:1140525-25-2 Formula:C24H29F3N4O6 IC50: 200 nM (C3a) Purity:98% SB290157 trifluoroacetate is a potent and selective C3a receptor antagonist with an IC 50 of 200 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.