Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SB290157 trifluoroacetate | 1140525-25-2 | MFCD04974198 | 99.8% | 526.51 g/mol | C24H29F3N4O6 | 10 MG
SDP

Catalog No. 5000245780
Encompass_Preferred

SB290157 trifluoroacetate is a selective antagonist of the complement C3a receptor (C3aR) used in research to probe C3a-mediated signaling. The compound is supplied as the trifluoroacetate salt, with reported in vitro potency (IC50 200 nM) and high chemical purity for laboratory studies.

  • Selective C3a receptor antagonist with reported IC50 of 200 nM.
  • Supplied as the trifluoroacetate salt.
  • Molecular formula C24H29F3N4O6; molecular weight 526.51 g/mol.
  • High purity (≈99.8%).
  • Available in milligram-scale packages for laboratory use.
  • For research use only; not for human or veterinary use.

Catalog No. 50-002-45780 Supplier Medchemexpress LLC Supplier No. HY101502A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SB290157 trifluoroacetate is a selective antagonist of the complement C3a receptor (C3aR) used in research to probe C3a-mediated signaling. The compound is supplied as the trifluoroacetate salt, with reported in vitro potency (IC50 200 nM) and high chemical purity for laboratory studies.

  • Selective C3a receptor antagonist with reported IC50 of 200 nM.
  • Supplied as the trifluoroacetate salt.
  • Molecular formula C24H29F3N4O6; molecular weight 526.51 g/mol.
  • High purity (≈99.8%).
  • Available in milligram-scale packages for laboratory use.
  • For research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.