Learn More
Medchemexpress LLC HY-103361 5mg Medchemexpress, SB297006 CAS:58816-69-6 Purity:>98%
SDP
Catalog No. 501968086
Encompass_Preferred
Medchemexpress, HY-103361 5mg SB297006 CAS:58816-69-6 SB297006 is a CCR3 antagonist, which significantly inhibits proliferation and neurosphere formation in CCL11-treated neural progenitor cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8086 Supplier Medchemexpress LLC Supplier No. HY1033615MG
Medchemexpress, HY-103361 5mg SB297006 CAS:58816-69-6 SB297006 is a CCR3 antagonist, which significantly inhibits proliferation and neurosphere formation in CCL11-treated neural progenitor cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.