Learn More
AdipoGen SB366791 (25 mg)
SDP
Supplier:  AdipoGen AGCR10034M025
Encompass_Preferred
Chemical. CAS: 472981-92-3. Formula: C16H14ClNO2. MW: 287.8. Potent and selective TRPV1 antagonist. Glutamatergic synaptic transmission inhibitor.
Catalog No. 50-205-3790