Learn More
Selleck Chemical LLC SB431542 S1067-200mg
SDP
Catalog No. 503108364
Encompass_Preferred
SB431542 is a potent and selective inhibitor of ALK5 with IC50 of 94 nM in a cell-free assay 100-fold more selective for ALK5 than p38 MAPK and other kinases
Catalog No. 50-310-8364 Supplier Selleck Chemical LLC Supplier No. S1067200MG
SB431542 is a potent and selective inhibitor of ALK5 with IC50 of 94 nM in a cell-free assay 100-fold more selective for ALK5 than p38 MAPK and other kinases