Learn More
STEMCELL Technologies SB431542 (Hydrate), Size: 10 mg
SDP
Supplier:  STEMCELL Technologies 72234
Encompass
Inhibit ALK4, ALK5, and ALK7 with SB431542 (Inman et al. Mol Pharmacol, 2002; Laping et al. Mol Pharmacol, 2002). This TGF-β/Activin/NODAL pathway inhibitor enhances the differentiation of cardiomyocytes from mouse and human PSCs (Kattman et al. Cell Stem Cell, 2011). In combination with various small molecules, SB431542 is used to increase the efficiency of reprogramming human somatic cells to iPS cells (Lin et al. Nat Methods, 2009), promote differentiation of neural progenitor cells from human PSCs (Chambers et al. Nat Biotechnol, 2009; Chambers et al. Nat Biotechnol, 2012), and modulate the direct lineage reprogramming of fibroblasts to mature neurons (Li et al. Cell Stem Cell, 2015). SB431542 is supplied as the hydrate form of the molecule with ≥ 98% purity; Molecular weight 384.4 g/mol; CAS number 301836-41-9. Visit STEMCELL.com for more information and related products.
Catalog No. NC1504022
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov