Learn More
Abcam SB431542, ALK inhibitor, 1MG
Catalog No. 502584663
Encompass_Preferred
MW 384.4 Da, Purity >99%. Potent and selective inhibitor of Activin Like Kinase. Inhibits ALK 4, 5 (IC₅₀ = 94 nM), and 7.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4663 Supplier Abcam Supplier No. AB1201631MG
MW 384.4 Da, Purity >99%. Potent and selective inhibitor of Activin Like Kinase. Inhibits ALK 4, 5 (IC₅₀ = 94 nM), and 7.
The product is subject to the following: Abcam Restricted Use Statement