Learn More
Electron Microscopy Sciences TEM-Specimen SB50 Grid Box
SDP
Supplier:  Electron Microscopy Sciences 7113501
Encompass_Preferred
TEM grid storage box for routine handling and long term storage of 50 standard size TEM grids. Ergonomic design incorporates several features that overcome the disadvantages associated with storage boxes of the more conventional ʽsliding cover' design. Unique number on the face and on one end.
Blue arrow at the 12 o'clock position indicates the park position for the cover when not in use. This is a firm grip "click" position and cannot be moved accidentally, preventing spillage. Clear cover can be rotated smoothly through 360 deg once the slight initial resistance of the park position has been overcome exposing a maximum of 2 or 3 diamond shaped slots at any one time. The 50 diamond shaped slots have an alphanumeric referencing system.
Each box is supplied with an index record card. Designed to be stacked. Base made of ABS-PHAT and cover of CAB.
Catalog No. 50-930-840