Learn More
Apexbio Technology LLC SB525334, 10mM (in 1mL DMSO). Cas: 356559-20-1 MFCD: MFCD11045307. (TGF-beta1) receptor inhibitor
SDP
Supplier:  Apexbio Technology LLC A56025.1
Encompass_Preferred
SB525334 is a potent inhibitor of transforming growth factor-beta1 (TGF-beta1) receptor, activin receptor-like kinase (ALK5). It specifically inhibits ALK5 with IC50 value of 14.3nM, which shows 4-fold higher potency against ALK4 [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4956