Learn More
Apexbio Technology LLC SB742457, 25mg. Cas: 607742-69-8 MFCD: MFCD12828872. 5-HT6 receptor antagonist,highly selective and high affinity
SDP
Supplier:  Apexbio Technology LLC B225625
Encompass_Preferred
SB742457 is a selective antagonist of 5-HT6 receptor with a pKi value of 9.63 [1].SB742457 has been found to be a highly selective 5-HT6 receptor antagonist with high affinity to human 5-HT6 receptor. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5335