Learn More
Abcam SB742457 (Intepirdine), 5-HT6 antagonist, 25MG
Catalog No. 502683370
Encompass_Preferred
MW 353.4 Da. Potent (pKi = 9.63) and highly selective 5-HT6 antagonist. Greater thsan 100-fold selective against other receptors. Reverses learning impairment in animal models.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-3370 Supplier Abcam Supplier No. AB26969525MG
MW 353.4 Da. Potent (pKi = 9.63) and highly selective 5-HT6 antagonist. Greater thsan 100-fold selective against other receptors. Reverses learning impairment in animal models.
The product is subject to the following: Abcam Restricted Use Statement