Learn More
Selleck Chemical LLC Pracinostat (SB939) 50mg 929016-96-6
SDP
Supplier:  Selleck Chemical LLC S151550MG
Encompass_Preferred
Pracinostat (SB939) is a potent pan-HDAC inhibitor with IC50 of 40-140 nM with exception for HDAC6. It has no activity against the class III isoenzyme SIRT I. Pracinostat (SB939) induces apoptosis in tumor cells. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-881-7