Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sbap (Succinimidyl 3-(Bromoacetamido)Propionate)

Catalog No. 80365050MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-365-050MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 80-365-050MG Supplier Sigma Aldrich Supplier No. 80365050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

SBAP is a sulfhydryl-reactive and amine-reactive heterobifunctional crosslinker. The reagent's NHS ester reacts with primary amines at pH 7-9 to form stable amide bonds, and the bromacetyl reacts with sulfhydryl groups at pH >7.5 to form stable thioether bonds This reagent is useful for preparing cyclic peptides and peptide conjugates because the spacer maintains peptide-like character in the crosslinked species.

Specifications

CAS 2462-32-0
Melting Point 156°C to 161°C
Molecular Formula C19H22NO3PS
Linear Formula C19H22NO3PS
Quantity 100 mg
Synonym (1S,4S,5S)-5-(4-methoxyphenyl)-2-tosyl-2-aza-5-phosphabicyclo[2.2.1]heptane
Molecular Weight (g/mol) 375.42 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.