Learn More
Medchemexpress LLC Sbc-110736 | 1629166-02-4 | MFCD30541539 | 99.6% | 413.51 g·mol⁻1 | C26H27N3O2 | 5 MG
SDP
Catalog No. 501967880
Encompass_Preferred
SBC-110736 is a small-molecule inhibitor of proprotein convertase subtilisin/kexin type 9 (PCSK9) described in patent WO2014150395A1. It has demonstrated cholesterol-lowering effects in preclinical mouse models and is supplied for research use as a solid powder or as a DMSO solution.
  • Inhibits PCSK9 activity in vitro and in vivo.
  • Shown to reduce total cholesterol in high-fat diet mouse models.
  • High purity: 99.6%.
  • Molecular formula C26H27N3O2; molecular weight 413.51 g·mol⁻1.
  • Supplied as milligram-scale solids and 10 mM solution in DMSO.
  • Stable when stored as powder at -20°C; solutions stable at -80°C for long-term storage.
  • Safety data sheet available for handling and hazard information.
Catalog No. 50-196-7880 Supplier Medchemexpress LLC Supplier No. HY1018325MG
SBC-110736 is a small-molecule inhibitor of proprotein convertase subtilisin/kexin type 9 (PCSK9) described in patent WO2014150395A1. It has demonstrated cholesterol-lowering effects in preclinical mouse models and is supplied for research use as a solid powder or as a DMSO solution.
  • Inhibits PCSK9 activity in vitro and in vivo.
  • Shown to reduce total cholesterol in high-fat diet mouse models.
  • High purity: 99.6%.
  • Molecular formula C26H27N3O2; molecular weight 413.51 g·mol⁻1.
  • Supplied as milligram-scale solids and 10 mM solution in DMSO.
  • Stable when stored as powder at -20°C; solutions stable at -80°C for long-term storage.
  • Safety data sheet available for handling and hazard information.