Learn More
AdipoGen SBD-F
SDP
Supplier:  AdipoGen CDXF0002M005
Encompass_Preferred
Chemical. CAS 84806-27-9. Formula C6H6FN3O4S. MW 235.19. Synthetic. Fluorescent probe for thiols widely used in pre-column labeling of biological thiols for HPLC.
Catalog No. 50-256-0955