Learn More
Selleck Chemical LLC SBE 13 HCl 10mg
SDP
Supplier:  Selleck Chemical LLC S772010MG
Encompass_Preferred
SBE 13 HCl is a potent and selective PLK1 inhibitor with IC50 of 200 pM, >4000-fold selectivity over Aurora A kinase, Plk2 and Plk3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3299