Learn More
SBE 13 hydrochloride, R&D Systems™

Description
Specifications
Specifications
| CAS | 1052532-15-6 |
| Molecular Formula | C24H28Cl2N2O4 |
| Quantity | 10 mg |
| Synonym | sbe13 hydrochloride, sbe 13 hydrochloride, sbe 13 hcl, n-4-6-chloro-3-pyridinyl methoxy-3-methoxyphenyl methyl-3,4-dimethoxybenzeneethanamine hydrochloride, n-4-6-chloropyridin-3-yl methoxy-3-methoxybenzyl-2-3,4-dimethoxyphenyl ethanamine, c24h27cln2o4.clh, sbe-13 hcl;sbe13 hcl, sbe 13 hydrochloride hplc, n-4-6-chloropyridin-3-yl methoxy-3-methoxyphenyl methyl-2-3,4-dimethoxyphenyl ethan-1-amine-hydrogen chloride 1/1 |
| InChI Key | QBGSVDJLQQXEGG-UHFFFAOYSA-N |
| SMILES | COC1=C(C=C(C=C1)CCNCC2=CC(=C(C=C2)OCC3=CN=C(C=C3)Cl)OC)OC.Cl |
| IUPAC Name | N-[[4-[(6-chloropyridin-3-yl)methoxy]-3-methoxyphenyl]methyl]-2-(3,4-dimethoxyphenyl)ethanamine;hydrochloride |
| Molecular Weight (g/mol) | 479.398 |
| PubChem CID | 11948807 |
| Formula Weight | 479.4 |
| Show More |
Safety and Handling
| Recommended Storage | Desiccate at Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.