Learn More
Cayman Chemical SBE 13hydrochlorIde 10mg
SDP
Catalog No. 502985121
Encompass_Preferred
A potent inhibitor of Plk1(IC50 = 0.2 nM) that targets the inactive conformation of the enzyme; exhibits no activity against aurora A kinase and less effectively inhibites Plk2 (IC50 > 66 µM) and Plk3 (IC50 = 875 nM); induces cell cycle arrest, reduces cell proliferation, and induces apoptosis
Catalog No. 50-298-5121 Supplier Cayman Chemical Supplier No. 2124110 MG
A potent inhibitor of Plk1(IC50 = 0.2 nM) that targets the inactive conformation of the enzyme; exhibits no activity against aurora A kinase and less effectively inhibites Plk2 (IC50 > 66 µM) and Plk3 (IC50 = 875 nM); induces cell cycle arrest, reduces cell proliferation, and induces apoptosis