Learn More
Medchemexpress LLC HY-17031 500mg , SBE-β-CD CAS:182410-00-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY17031500MG
Encompass_Preferred
HY-17031 500mg SBE-β-CD CAS: 182410-00-0 SBE-β-CD is a sulfobutylether β-cyclodextrin derivative used as an excipient or a formulating agent to increase the solubility of poorly soluble agents. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-8989