Learn More
Medchemexpress LLC HY-17031 50g Medchemexpress, SBE-β-CD Sulfobutylether-β-Cyclodextrin CAS:182410-00-0 Purity:98%
SDP
Catalog No. 501638597
Encompass_Preferred
Medchemexpress,HY-17031 50g SBE-β-CD Sulfobutylether-β-Cyclodextrin CAS:182410-00-0 Formula:N/A SBE-β-CD is a sulfobutylether β-cyclodextrin derivative used as an excipient or a formulating agent to increase the solubility of poorly soluble drugs. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-8597 Supplier Medchemexpress LLC Supplier No. HY1703150G
Medchemexpress,HY-17031 50g SBE-β-CD Sulfobutylether-β-Cyclodextrin CAS:182410-00-0 Formula:N/A SBE-β-CD is a sulfobutylether β-cyclodextrin derivative used as an excipient or a formulating agent to increase the solubility of poorly soluble drugs. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.