Learn More
Medchemexpress LLC HY-17031 5g Medchemexpress, SBE-β-CD CAS:182410-00-0 Purity:>98%
SDP
Catalog No. 502029230
Encompass_Preferred
Medchemexpress, HY-17031 5g SBE-β-CD CAS:182410-00-0 SBE-β-CD is a sulfobutylether β-cyclodextrin derivative used as an excipient or a formulating agent to increase the solubility of poorly soluble agents. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9230 Supplier Medchemexpress LLC Supplier No. HY170315G
Medchemexpress, HY-17031 5g SBE-β-CD CAS:182410-00-0 SBE-β-CD is a sulfobutylether β-cyclodextrin derivative used as an excipient or a formulating agent to increase the solubility of poorly soluble agents. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.