Learn More
BPS Bioscience Inc SBE Luciferase Reporter Lentivirus, 2 vials, 500 μl each, TGFβ/SMAD signaling pathway, Firefly Luciferase, ≥5 x 10^6 TU/ml, -80°C storage
SDP
Catalog No. NC3640435
Encompass
The SBE Luciferase Reporter Lentivirus is engineered to facilitate the monitoring of the TGFβ/SMAD signaling pathway. This replication-incompetent lentiviral particle, utilizing HIV-based and VSV-G pseudotyping technology, incorporates a firefly luciferase gene regulated by a multimerized SBE-responsive element. It offers a sensitive method for detecting changes in the TGFβ/SMAD pathway activity in mammalian cells. This product serves as an essential tool for those aiming to screen potential pathway modulators and develop stable reporter cell lines.
Catalog No. NC3640435 Supplier BPS Bioscience Inc Supplier No. 79806
The SBE Luciferase Reporter Lentivirus is engineered to facilitate the monitoring of the TGFβ/SMAD signaling pathway. This replication-incompetent lentiviral particle, utilizing HIV-based and VSV-G pseudotyping technology, incorporates a firefly luciferase gene regulated by a multimerized SBE-responsive element. It offers a sensitive method for detecting changes in the TGFβ/SMAD pathway activity in mammalian cells. This product serves as an essential tool for those aiming to screen potential pathway modulators and develop stable reporter cell lines.