Learn More
Medchemexpress LLC SBE13 Hydrochloride | 1052532-15-6 | C24H28Cl2N2O4 | 1 ML
SDP
Catalog No. 5000373991
Encompass_Preferred
SBE13 Hydrochloride is a potent and selective Plk1 inhibitor, with an IC50 of 200 pM. It poorly inhibits Plk2 (IC50>66 μM) or Plk3 (IC50=875 nM). This compound is for research use only.
  • Potent and selective Plk1 inhibitor
  • Induces apoptosis in HeLa cells
  • Reduces cell proliferation
  • Can be stored at 4°C for solid form, and -80°C or -20°C in solvent
Catalog No. 50-003-73991 Supplier Medchemexpress LLC Supplier No. HY1515810MM/1ML
SBE13 Hydrochloride is a potent and selective Plk1 inhibitor, with an IC50 of 200 pM. It poorly inhibits Plk2 (IC50>66 μM) or Plk3 (IC50=875 nM). This compound is for research use only.
  • Potent and selective Plk1 inhibitor
  • Induces apoptosis in HeLa cells
  • Reduces cell proliferation
  • Can be stored at 4°C for solid form, and -80°C or -20°C in solvent