Learn More
TARGETMOL CHEMICALS INC SBE13 HYDROCHLORIDE 25MG
SDP
Catalog No. 502580931
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SBE13 Hydrochloride (SBE 13 HCl) is an effective and specific PLK1 inhibitor (IC50 0.2 nM); no inhibition om Aurora A kinase Plk2/3. purity: 99%
Catalog No. 50-258-0931 Supplier TARGETMOL CHEMICALS INC Supplier No. T227125MG
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SBE13 Hydrochloride (SBE 13 HCl) is an effective and specific PLK1 inhibitor (IC50 0.2 nM); no inhibition om Aurora A kinase Plk2/3. purity: 99%