Learn More
LGC GENOMICS LLC sbeade pathogen nucleic acid purification kit (96 purifications)
Catalog No. NC3920029
Encompass
Key features:Rapid nucleic acid purification from various pathogensIncludes Proteinase K to facilitate efficient cell disruption and nucleic acid releaseHigh-quality sbeadex magnetic bead-based technologyReady-to-use kit compatible with most robotic platformsSuitable for most downstream applications such as: PCR/qPCR, RT-PCR/qRT-PCR, and NGS
Catalog No. NC3920029 Supplier LGC GENOMICS LLC Supplier No. NAP4002401
Add to Cart
Add to Cart
Key features:Rapid nucleic acid purification from various pathogensIncludes Proteinase K to facilitate efficient cell disruption and nucleic acid releaseHigh-quality sbeadex magnetic bead-based technologyReady-to-use kit compatible with most robotic platformsSuitable for most downstream applications such as: PCR/qPCR, RT-PCR/qRT-PCR, and NGS