Learn More
LGC GENOMICS LLC sbeadex Livestock DNA Purification Kit - 96 purifications
Catalog No. NC3920027
Encompass
Key FeaturesUniversal solution for DNA purification from diverse livestock samplesSuitable for various downstream applications, including next-generation sequencing, KASP genotyping, and Chip technologiesSimplified workflow with standard protocol that eliminates the need for multiple kits specific for different sample typesCompatible with popular robotic platforms like KingFisher, Hamilton, or Tecan instruments, allowing for efficient high-throughput extractions and easy automation
Catalog No. NC3920027 Supplier LGC GENOMICS LLC Supplier No. NAP44701
Key FeaturesUniversal solution for DNA purification from diverse livestock samplesSuitable for various downstream applications, including next-generation sequencing, KASP genotyping, and Chip technologiesSimplified workflow with standard protocol that eliminates the need for multiple kits specific for different sample typesCompatible with popular robotic platforms like KingFisher, Hamilton, or Tecan instruments, allowing for efficient high-throughput extractions and easy automation