Learn More
LGC GENOMICS LLC (Lucigen) sbeadex™ SAB (sequencing application beads), 20 mLHighly cost-effective alternative to AMPure XP beads in PCR and NGS clean-up and size selection applications.
Catalog No. 502231957
Encompass
Highly cost-effective magnetic beads for PCR and NGS clean-up as well as size selection applications.
This item is not returnable. View return policy
Catalog No. 50-223-1957 Supplier LGC GENOMICS LLC Supplier No. NAP45002
Highly cost-effective magnetic beads for PCR and NGS clean-up as well as size selection applications.