Learn More
Apexbio Technology LLC SBI-0206965, 1884220-36-3, MFCD29472244, 5 mg
SDP
Supplier:  Apexbio Technology LLC A87155
Encompass_Preferred
MF: C21H21BrN4O5, MW: 489.32, Solubility: 24.5mg/mL in DMSO. SBI-0206965 is a novel inhibitor of the autophagy-initiating kinase ULK1.
Catalog No. 50-115-1126