Learn More
Medchemexpress LLC HY-111534 10mg Medchemexpress, SBI-115 CAS:882366-16-7 Purity:>98%
SDP
Catalog No. 501968903
Encompass_Preferred
Medchemexpress, HY-111534 10mg SBI-115 CAS:882366-16-7 SBI-115 is a TGR5 (GPCR19) antagonist. SBI-115 decreases hepatic cystogenesis with polycystic liver diseases via inhibiting TGR5. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8903 Supplier Medchemexpress LLC Supplier No. HY11153410MG
Medchemexpress, HY-111534 10mg SBI-115 CAS:882366-16-7 SBI-115 is a TGR5 (GPCR19) antagonist. SBI-115 decreases hepatic cystogenesis with polycystic liver diseases via inhibiting TGR5. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.