Learn More
Selleck Chemical LLC SBI-115
SDP
Catalog No. 5000002850
Encompass_Preferred
SBI-115 is a novel TGR5 (GPCR19) antagonist SBI-115 decreases cell proliferation cholangiocyte spheroid growth and cAMP levels in cystic cholangiocytes
Catalog No. 50-000-02850 Supplier Selleck Chemical LLC Supplier No. S68062MG
SBI-115 is a novel TGR5 (GPCR19) antagonist SBI-115 decreases cell proliferation cholangiocyte spheroid growth and cAMP levels in cystic cholangiocytes