Learn More
Medchemexpress LLC HY-124756 5mg Medchemexpress, SBI-425 CAS:1451272-71-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1247565MG
Encompass_Preferred
Medchemexpress, HY-124756 5mg SBI-425 CAS:1451272-71-1 SBI-425 is a potent, selective and oral bioavailable tissue-nonspecific alkaline phosphatase (TNAP) inhibitor. Long-term administration of SBI-425 effectively reaches and inhibits TNAP in the vasculature, improving cardiovascular parameters and survival at a dose that does not cause a detectable change in bone. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9681