Learn More
Medchemexpress LLC HY-124418 1mg Medchemexpress, SBI-477 CAS:781628-99-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1244181MG
Encompass_Preferred
Medchemexpress, HY-124418 1mg SBI-477 CAS:781628-99-7 SBI-477 is a chemical probe stimulated insulin signaling by deactivating the transcription factor MondoA, leading to reduced expression of the insulin pathway suppressors thioredoxin-interacting protein (TXNIP) and arrestin domain–containing 4 (ARRDC4). SBI-477 coordinately inhibits triacylglyceride (TAG) synthesis and enhances basal glucose uptake in human skeletal myocytes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9663