Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sbi-477 | 781628-99-7 | MFCD04508832 | 98.0% | 483.54 | C24H25N3O6S | 5MG
SDP

Catalog No. 5000239149
Encompass_Preferred

SBI-477 is a small-molecule chemical probe that deactivates the transcription factor MondoA, stimulating insulin signaling and reducing expression of insulin-pathway suppressors such as TXNIP. It is supplied as a purified solid intended for research use and is characterized by molecular weight 483.54 and formula C24H25N3O6S.

  • Deactivates MondoA to stimulate insulin signaling.
  • Used in cellular studies to modulate glucose uptake and lipid synthesis.
  • Purity 98.0%.
  • Molecular weight 483.54 g/mol.
  • Chemical formula C24H25N3O6S.
  • Soluble in DMSO (≈65 mg/mL); hygroscopic-use fresh DMSO and ultrasonic assistance if needed.
  • Store powder at -20°C for long-term stability; follow solvent storage recommendations for solutions.

Catalog No. 50-002-39149 Supplier Medchemexpress LLC Supplier No. HY1244185MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SBI-477 is a small-molecule chemical probe that deactivates the transcription factor MondoA, stimulating insulin signaling and reducing expression of insulin-pathway suppressors such as TXNIP. It is supplied as a purified solid intended for research use and is characterized by molecular weight 483.54 and formula C24H25N3O6S.

  • Deactivates MondoA to stimulate insulin signaling.
  • Used in cellular studies to modulate glucose uptake and lipid synthesis.
  • Purity 98.0%.
  • Molecular weight 483.54 g/mol.
  • Chemical formula C24H25N3O6S.
  • Soluble in DMSO (≈65 mg/mL); hygroscopic-use fresh DMSO and ultrasonic assistance if needed.
  • Store powder at -20°C for long-term stability; follow solvent storage recommendations for solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.