Learn More
Medchemexpress LLC HY-125880 5mg Medchemexpress, SBI-553 CAS:1849603-72-0 Purity:>98%
SDP
Catalog No. 501969712
Encompass_Preferred
Medchemexpress, HY-125880 5mg SBI-553 CAS:1849603-72-0 SBI-553 is a potent and brain penetrant NTR1 allosteric modulator, with an EC50 of 0.34 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9712 Supplier Medchemexpress LLC Supplier No. HY1258805MG
Medchemexpress, HY-125880 5mg SBI-553 CAS:1849603-72-0 SBI-553 is a potent and brain penetrant NTR1 allosteric modulator, with an EC50 of 0.34 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.