Learn More
Medchemexpress LLC SBI-581 | 98.2% | 383.44 | 1 ML
SDP
Catalog No. 5000351658
Encompass_Preferred
SBI-581 is an orally active and potent selective serine-threonine kinase TAO3 inhibitor with an IC50 of 42 nM. It promotes TKS5α accumulation at RAB11-positive vesicles, inhibits invadopodia formation, and shows reasonable pharmacokinetics and antitumor activity in mice. It is also a click chemistry reagent with an Alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc).
  • Promotes TKS5α accumulation at RAB11-positive vesicles
  • Inhibits invadopodia formation
  • Shows reasonable pharmacokinetics and antitumor activity in mice
  • Functions as a click chemistry reagent with an Alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Displays moderate selectivity against a broad panel of kinases
Catalog No. 50-003-51658 Supplier Medchemexpress LLC Supplier No. HY13943910MM/1ML
SBI-581 is an orally active and potent selective serine-threonine kinase TAO3 inhibitor with an IC50 of 42 nM. It promotes TKS5α accumulation at RAB11-positive vesicles, inhibits invadopodia formation, and shows reasonable pharmacokinetics and antitumor activity in mice. It is also a click chemistry reagent with an Alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc).
  • Promotes TKS5α accumulation at RAB11-positive vesicles
  • Inhibits invadopodia formation
  • Shows reasonable pharmacokinetics and antitumor activity in mice
  • Functions as a click chemistry reagent with an Alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Displays moderate selectivity against a broad panel of kinases