Learn More
Medchemexpress LLC SBI-581 | 98.2% | 383.44 | 25 MG
SDP
Catalog No. 5000348158
Encompass_Preferred
SBI-581 is an orally active and potent selective serine-threonine kinase TAO3 inhibitor with an IC50 of 42 nM. It promotes TKS5α accumulation at RAB11-positive vesicles and inhibits invadopodia formation. The compound demonstrates reasonable pharmacokinetics in mice and shows antitumor activity. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Orally active and potent selective serine-threonine kinase TAO3 inhibitor
  • Promotes TKS5α accumulation at RAB11-positive vesicles
  • Inhibits invadopodia formation
  • Demonstrates reasonable pharmacokinetics in mice
  • Shows antitumor activity
  • Click chemistry reagent for copper-catalyzed azide-alkyne cycloaddition
  • Moderate selectivity against a broad panel of kinases
Catalog No. 50-003-48158 Supplier Medchemexpress LLC Supplier No. 25MGHY139439
SBI-581 is an orally active and potent selective serine-threonine kinase TAO3 inhibitor with an IC50 of 42 nM. It promotes TKS5α accumulation at RAB11-positive vesicles and inhibits invadopodia formation. The compound demonstrates reasonable pharmacokinetics in mice and shows antitumor activity. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Orally active and potent selective serine-threonine kinase TAO3 inhibitor
  • Promotes TKS5α accumulation at RAB11-positive vesicles
  • Inhibits invadopodia formation
  • Demonstrates reasonable pharmacokinetics in mice
  • Shows antitumor activity
  • Click chemistry reagent for copper-catalyzed azide-alkyne cycloaddition
  • Moderate selectivity against a broad panel of kinases