Learn More
Selleck Chemical LLC SBI-797812 5mg
SDP
Supplier:  Selleck Chemical LLC S89805MG
Encompass_Preferred
SBI-797812 is a NAMPT activator which is structurally similar to active-site directed NAMPT inhibitors and blocks binding of these inhibitors to NAMPT. SBI-797812 turns NAMPT into a "super catalyst" that more efficiently generates NMN. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3464