Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SBI-810 hydrochloride | 2772746-58-2 | 99.6% | 483.05 | C27H35ClN4O2 | 10 MG
SDP

Catalog No. 5000289294
Encompass_Preferred

SBI-810 hydrochloride is a β-arrestin-biased allosteric modulator of the neurotensin receptor 1 (NTSR1) supplied as the hydrochloride salt for research use. Provided as a solid with high analytical purity, it is intended for in vitro pharmacology and preclinical signaling or analgesia studies.

  • β-arrestin-biased NTSR1 allosteric modulator for receptor signaling studies.
  • High purity; typically ≥99.5% by supplier assay.
  • Molecular weight 483.05; formula C27H35ClN4O2.
  • Available in small-mass pack sizes suitable for in vitro and preclinical work.
  • Option for 10 mM solution in DMSO, ready for reconstitution.
  • Store sealed at 4°C; in solution store at -80°C for long-term storage.

Catalog No. 50-002-89294 Supplier Medchemexpress LLC Supplier No. HY164795A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SBI-810 hydrochloride is a β-arrestin-biased allosteric modulator of the neurotensin receptor 1 (NTSR1) supplied as the hydrochloride salt for research use. Provided as a solid with high analytical purity, it is intended for in vitro pharmacology and preclinical signaling or analgesia studies.

  • β-arrestin-biased NTSR1 allosteric modulator for receptor signaling studies.
  • High purity; typically ≥99.5% by supplier assay.
  • Molecular weight 483.05; formula C27H35ClN4O2.
  • Available in small-mass pack sizes suitable for in vitro and preclinical work.
  • Option for 10 mM solution in DMSO, ready for reconstitution.
  • Store sealed at 4°C; in solution store at -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.