Learn More
Aviva Systems Biology Corp Sbk1 Antibody - C-terminal region (ARP48523_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48523_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Sbk1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: SbkApplicationWB. Species reactivity: Guinea Pig, Human, Mouse, Rat
Catalog No. 50-285-365