Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human SBK1, GST Tag Recombinant Protein

Catalog No. A30518
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30518 10 μg
A32877 100 μg
A33367 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A30518 Supplier Invitrogen™ Supplier No. A30518
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This SBK1 recombinant human protein (full length) was expressed in insect cells. SBK1 recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Z'-LYTE™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Full Length, Recombinant, Active The gene accession number for SBK1 is NP_001019572.1.

SBK1 novel serine/threonine kinase or SH3 domain binding kinase 1 is structurally related to a Xenopus gastrula-specific protein kinase, Pk9.7 which possessing a consensus sequence for an SH3-binding domain from developing rat brain. SBK1 plays an important role in signal-transduction pathways related to the control of brain development. SBK1 may predict a broad cellular function, and its dysregulated in certain cancers suggests an involvement of the protein in the pathogenesis of human cancers. May be involved in signal-transduction pathways related to the control of brain development. Target protein is a serine/threonine protein kinase gene expressed predominantly in developing brain.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q52WX2
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 10mM glutathione, 150mM NaCl, 0.1mM PMSF, 0.1mM EDTA, 25% glycerol, 0.25mM DTT and no preservative; pH 7.5
Gene ID (Entrez) 388228
Molecular Weight (g/mol) 72.5 kDa
Name Human SBK1, GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias SBK; SBK1; Serine/threonine-protein kinase SBK1; SH3 domain binding kinase 1; SH3-binding domain kinase 1; SH3-binding kinase 1
Common Name Sbk1
Gene Symbol SBK1
Kinase Family NKF1 Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Form Full Length, Recombinant, Active
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.