Learn More
Medchemexpress LLC SBP-0636457 | 1422180-49-1 | 98.4% | 456.58 | C25H36N4O4 | 10MG
SDP
Catalog No. 5000235527
Encompass_Preferred
SBP-0636457 is a small-molecule SMAC mimetic that antagonizes inhibitor of apoptosis proteins (IAPs). It binds the BIR domains of IAP family proteins (reported Ki ≈ 0.27 μM) and is used as a research tool in apoptosis and oncology studies. The compound is supplied as a high-purity solid and is intended for laboratory research applications.
Catalog No. 50-002-35527 Supplier Medchemexpress LLC Supplier No. HY12537810MG
SBP-0636457 is a small-molecule SMAC mimetic that antagonizes inhibitor of apoptosis proteins (IAPs). It binds the BIR domains of IAP family proteins (reported Ki ≈ 0.27 μM) and is used as a research tool in apoptosis and oncology studies. The compound is supplied as a high-purity solid and is intended for laboratory research applications.