Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SBP-3264 | 2869148-13-8 | 98.5% | C19H20ClN5O | 10MG
SDP

Supplier:  Medchemexpress LLC HY13296910MG

Encompass_Preferred

SBP-3264 is a fluorescent small-molecule chemical probe used to investigate STK3 and STK4 activity and Hippo pathway signaling in acute myeloid leukemia (AML). It is supplied as a high-purity solid with defined solubility in DMSO and recommended storage conditions to preserve stability for in vitro and cellular studies.

  • High purity: 98.5% by HPLC.
  • Molecular formula C19H20ClN5O; molecular weight 369.85 g/mol.
  • Soluble in DMSO at 62.5 mg/mL (≈169 mM) for preparation of stock solutions.
  • Suitable for in vitro and cellular assays targeting Hippo pathway kinases.
  • Recommended storage: protect from light; store dry at 4°C, in solution -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-58859


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.