Learn More
Selleck Chemical LLC SBP-3264-E1065-25MG
SDP
Catalog No. 503137215
Encompass_Preferred
SBP-3264 is a valuable chemical probe for understanding the roles of STK3 and STK4 in acute myeloid leukemia (AML)
Catalog No. 50-313-7215 Supplier Selleck Chemical LLC Supplier No. E106525MG
SBP-3264 is a valuable chemical probe for understanding the roles of STK3 and STK4 in acute myeloid leukemia (AML)