Learn More
Medchemexpress LLC HY-137742 5mg Medchemexpress, SBP-7455 CAS:1884222-74-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1377425MG
Encompass_Preferred
Medchemexpress, HY-137742 5mg SBP-7455 CAS:1884222-74-5 SBP-7455 is a potent, high affinity and orally active dual ULK1/ULK2 autophagy inhibitor with IC50s of 13 nM and 476 nM in the ADP-Glo assays, respectively. SBP-7455 potently inhibits ULK1/2 enzymatic activity and can be used for triple-negative breast cancer (TNBC) research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1507