Learn More
Cayman Chemical SBP1trIfluoroacetate salt 10mg
SDP
Catalog No. 502915594
Encompass_Preferred
A peptide; LNPs encapsulating oseltamivir phosphate and conjugated to SBP1 have been studied in vitro to estimate their potential for providing for controlled, long-term release of oseltamivir phosphate in vivo; increases network capture efficiency of SARS-CoV-2 spike glycoprotein in vitro when immobilized to crosslinked networks of hydroxy ethyl acrylate copolymers at 2%
Catalog No. 50-291-5594 Supplier Cayman Chemical Supplier No. 3746910 MG
A peptide; LNPs encapsulating oseltamivir phosphate and conjugated to SBP1 have been studied in vitro to estimate their potential for providing for controlled, long-term release of oseltamivir phosphate in vivo; increases network capture efficiency of SARS-CoV-2 spike glycoprotein in vitro when immobilized to crosslinked networks of hydroxy ethyl acrylate copolymers at 2%