Learn More
Cayman Chemical (S)-Bromoenol lactone, Purity: ≥98%, CAS: 478288-94-7, 5 mg
SDP
Catalog No. 502889934
Encompass_Preferred
A selective iPLA2β inhibitor; inhibits vasopressin-induced release of arachidonate from cultured rat aortic smooth muscle (A10) cells (IC50 = 2 µM); >1,000-fold selective for iPLA2 versus cPLA2 and 10-fold selective for iPLA2β versus iPLA2γ
This item is not returnable. View return policy
Catalog No. 50-288-9934 Supplier Cayman Chemical Supplier No. 100068015 MG
A selective iPLA2β inhibitor; inhibits vasopressin-induced release of arachidonate from cultured rat aortic smooth muscle (A10) cells (IC50 = 2 µM); >1,000-fold selective for iPLA2 versus cPLA2 and 10-fold selective for iPLA2β versus iPLA2γ