Learn More
Cayman Chemical SBromoenol lactone
SDP
Supplier:  Cayman Chemical 10006801500 UG
Encompass_Preferred
A selective iPLA2β inhibitor; inhibits vasopressin-induced release of arachidonate from cultured rat aortic smooth muscle (A10) cells (IC50 = 2 µM); >1,000-fold selective for iPLA2 versus cPLA2 and 10-fold selective for iPLA2β versus iPLA2γ
Catalog No. 50-288-9170