Learn More
Cayman Chemical (S)-Bromoenol lactone, Purity: ≥98%, CAS: 478288-94-7, 10 mg
SDP
Catalog No. 502890961
Encompass_Preferred
A selective iPLA2β inhibitor; inhibits vasopressin-induced release of arachidonate from cultured rat aortic smooth muscle (A10) cells (IC50 = 2 µM); >1,000-fold selective for iPLA2 versus cPLA2 and 10-fold selective for iPLA2β versus iPLA2γ
Catalog No. 50-289-0961 Supplier Cayman Chemical Supplier No. 1000680110 MG
A selective iPLA2β inhibitor; inhibits vasopressin-induced release of arachidonate from cultured rat aortic smooth muscle (A10) cells (IC50 = 2 µM); >1,000-fold selective for iPLA2 versus cPLA2 and 10-fold selective for iPLA2β versus iPLA2γ