Learn More
SPEX CERTIPREP LLC 3P Scandium (10,000g/mL in 4% HNO3), 100mL
Catalog No. NC1647711
Encompass_Preferred
(HPS by Zeptometrix) 10,000 g/mL Scandium in 4% HNO. This standard is traceable to NIST SRM 3148 and certified ISO 17034 and 17025.
Catalog No. NC1647711 Supplier SPEX CERTIPREP LLC 3P Supplier No. 10M481100
(HPS by Zeptometrix) 10,000 g/mL Scandium in 4% HNO. This standard is traceable to NIST SRM 3148 and certified ISO 17034 and 17025.